Assay Details
| Gene Symbol: GTF2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038064.1 | NP_001033153.1 | 5 - 6 | 442 | 78 | |
| GenBank mRNA | BC111360.1 | - | 5 - 6 | 442 | 78 | |
More Information
Set Membership: |
|

GTF2F2
Genomic Map