Assay Details
| Gene Symbol: ARL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038079.2 | NP_001033168.1 | 4 - 5 | 716 | 86 | |
| GenBank mRNA | BC111133.1 | - | 4 - 5 | 717 | 86 | |
More Information
Set Membership: |
|

ARL2
Genomic Map