Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038116.2 | NP_001033205.2 | 1 - 2 | 265 | 56 | |
| GenBank mRNA | BC109701.1 | - | 1 - 2 | 247 | 56 | |
More Information
Set Membership: |
|

PIGH
Genomic Map