Assay Details
| Gene Symbol: GMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038119.1 | NP_001033208.1 | 3 - 4 | 281 | 70 | |
| GenBank mRNA | BC109695.1 | - | 3 - 4 | 281 | 70 | |
More Information
Set Membership: |
|

GMPR2
Genomic Map