Assay Details
| Gene Symbol: CDKL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038129.2 | NP_001033218.1 | 7 - 8 | 841 | 124 | |
| GenBank mRNA | BC111325.1 | - | 7 - 8 | 842 | 124 | |
More Information
Set Membership: |
|

CDKL4
Genomic Map