Assay Details
| Gene Symbol: EFCAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038141.2 | NP_001033230.1 | 5 - 6 | 594 | 75 | |
| GenBank mRNA | BC111336.1 | - | 5 - 6 | 595 | 75 | |
More Information
Set Membership: |
|

EFCAB3
Genomic Map