Assay Details
| Gene Symbol: PRCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038164.2 | NP_001033253.1 | 4 - 5 | 476 | 89 | |
| GenBank mRNA | BC111171.1 | - | 4 - 5 | 477 | 89 | |
More Information
Set Membership: |
|

PRCP
Genomic Map