Assay Details
Gene Symbol: TRIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038165.2 | NP_001033254.1 | 2 - 3 | 389 | 135 | |
GenBank mRNA | BC109749.1 | - | 2 - 3 | 390 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109749.1, NM_001038165.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
