Assay Details
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038170.2 | NP_001033259.1 | 9 - 10 | 1262 | 93 | |
GenBank mRNA | BC109933.1 | - | 9 - 10 | 1263 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
