Assay Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038182.2 | NP_001033271.1 | 8 - 9 | 1061 | 64 | |
| GenBank mRNA | BC109620.1 | - | 8 - 9 | 1062 | 64 | |
| EF534775.1 | - | 8 - 9 | 1052 | 64 | ||
More Information
Set Membership: |
|

DMRTC2
Genomic Map