Assay Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038185.2 | NP_001033274.1 | 5 - 6 | 800 | 70 | |
| GenBank mRNA | BC109758.1 | - | 5 - 6 | 801 | 70 | |
More Information
Set Membership: |
|

SPAG6
Genomic Map