Assay Details
| Gene Symbol: H2AZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038197.1 | NP_001033286.1 | 4 - 5 | 384 | 86 | |
| GenBank mRNA | BC109673.1 | - | 4 - 5 | 423 | 86 | |
More Information
Set Membership: |
|

H2AZ2
Genomic Map