Assay Details
Gene Symbol: PRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038200.1 | NP_001033289.1 | 3 - 4 | 390 | 113 | |
GenBank mRNA | AB196438.1 | - | 3 - 4 | 390 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
