Assay Details
| Gene Symbol: NFIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038209.2 | NP_001033298.1 | 5 - 6 | 902 | 66 | |
| GenBank mRNA | BC109482.1 | - | 5 - 6 | 903 | 66 | |
More Information
Set Membership: |
|

NFIA
Genomic Map