Assay Details
| Gene Symbol: TCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038211.1 | NP_001033300.1 | 4 - 5 | 788 | 69 | |
| GenBank mRNA | BC111345.1 | - | 4 - 5 | 788 | 69 | |
More Information
Set Membership: |
|

TCAM1
Genomic Map