Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038501.2 | NP_001033590.1 | 2 - 3 | 114 | 127 | |
GenBank mRNA | BC110212.1 | - | 2 - 3 | 115 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110212.1, NM_001038501.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
