Assay Details
| Gene Symbol: DDX25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038517.1 | NP_001033606.1 | 9 - 10 | 1049 | 65 | |
| GenBank mRNA | BC109867.1 | - | 9 - 10 | 1049 | 65 | |
More Information
Set Membership: |
|

DDX25
Genomic Map