Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038571.2 | NP_001033660.1 | 3 - 4 | 2328 | 68 | |
| GenBank mRNA | BC149048.1 | - | 4 - 5 | 2105 | 68 | |
| BT021194.1 | - | 1 - 2 | 2109 | 68 | ||
| DY461183.1 | - | 1 - 2 | 67 | 68 | ||
More Information
Set Membership: |
|

GPR61
Genomic Map