Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038647.1 | NP_001033736.1 | 10 - 11 | 1916 | 58 | |
| GenBank mRNA | AY845018.1 | - | 10 - 11 | 1916 | 58 | |
More Information
Set Membership: |
|

MACO1
Genomic Map