Assay Details
| Gene Symbol: SAA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040505.2 | NP_001035595.1 | 3 - 4 | 393 | 69 | |
| GenBank mRNA | BC109725.1 | - | 3 - 4 | 394 | 69 | |
More Information
Set Membership: |
|

SAA4
Genomic Map