Assay Details
| Gene Symbol: SAA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040505.2 | NP_001035595.1 | 4 - 5 | 532 | 82 | |
| GenBank mRNA | BC109725.1 | - | 4 - 5 | 533 | 82 | |
More Information
Set Membership: |
|

SAA4
Genomic Map