Assay Details
| Gene Symbol: RPL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040520.2 | NP_001035610.1 | 2 - 3 | 151 | 101 | |
| GenBank mRNA | BC109810.1 | - | 2 - 3 | 152 | 101 | |
| BT030549.1 | - | 1 - 2 | 127 | 101 | ||
More Information
Set Membership: |
|

RPL7A
Genomic Map