Assay Details
Gene Symbol: CAPZA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040561.2 | NP_001035651.1 | 1 | 948 | 86 | |
GenBank mRNA | BC109951.1 | - | 1 | 949 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
