Assay Details
| Gene Symbol: BRI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040583.1 | NP_001035673.1 | 3 - 4 | 420 | 86 | |
| GenBank mRNA | BC109710.1 | - | 3 - 4 | 420 | 86 | |
More Information
Set Membership: |
|

BRI3
Genomic Map