Assay Details
| Gene Symbol: TRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045896.2 | NP_001039361.1 | 1 - 2 | 70 | 77 | |
| GenBank mRNA | BC112743.1 | - | 1 - 2 | 71 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112743.1, NM_001045896.2 transcripts.
Set Membership: |
|

TRADD
Genomic Map