Assay Details
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045902.1 | NP_001039367.1 | 1 - 2 | 142 | 98 | |
GenBank mRNA | BC112487.1 | - | 1 - 2 | 142 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
