Assay Details
| Gene Symbol: CD68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045902.1 | NP_001039367.1 | 4 - 5 | 803 | 72 | |
| GenBank mRNA | BC112487.1 | - | 4 - 5 | 803 | 72 | |
More Information
Set Membership: |
|

CD68
Genomic Map