Assay Details
| Gene Symbol: RRAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045913.1 | NP_001039378.1 | 1 - 2 | 114 | 73 | |
| GenBank mRNA | BC112722.1 | - | 1 - 2 | 114 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112722.1, NM_001045913.1 transcripts.
Set Membership: |
|

RRAD
Genomic Map