Assay Details
| Gene Symbol: RRAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045913.1 | NP_001039378.1 | 2 - 3 | 503 | 72 | |
| GenBank mRNA | BC112722.1 | - | 2 - 3 | 503 | 72 | |
More Information
Set Membership: |
|

RRAD
Genomic Map