Assay Details
| Gene Symbol: APOA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045916.2 | NP_001039381.1 | 3 - 4 | 243 | 59 | |
| GenBank mRNA | BC111607.1 | - | 3 - 4 | 245 | 59 | |
More Information
Set Membership: |
|

APOA2
Genomic Map