Assay Details
| Gene Symbol: MPV17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045929.2 | NP_001039394.1 | 7 - 8 | 528 | 70 | |
| GenBank mRNA | BC112572.1 | - | 7 - 8 | 529 | 70 | |
More Information
Set Membership: |
|

MPV17
Genomic Map