Assay Details
| Gene Symbol: DDX55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046007.2 | NP_001039472.1 | 3 - 4 | 290 | 62 | |
| GenBank mRNA | BC111255.1 | - | 3 - 4 | 291 | 62 | |
More Information
Set Membership: |
|

DDX55
Genomic Map