Assay Details
| Gene Symbol: ASPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046033.2 | NP_001039498.1 | 2 - 3 | 326 | 90 | |
| GenBank mRNA | BC112799.1 | - | 2 - 3 | 327 | 90 | |
More Information
Set Membership: |
|

ASPA
Genomic Map