Assay Details
| Gene Symbol: CYRIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046047.2 | NP_001039512.1 | 12 - 13 | 1092 | 85 | |
| GenBank mRNA | BC105327.1 | - | 12 - 13 | 1094 | 85 | |
More Information
Set Membership: |
|

CYRIB
Genomic Map