Assay Details
| Gene Symbol: RPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046083.2 | NP_001039548.1 | 8 - 9 | 679 | 74 | |
| GenBank mRNA | BC110138.1 | - | 8 - 9 | 680 | 74 | |
More Information
Set Membership: |
|

RPF2
Genomic Map