Assay Details
Gene Symbol: COQ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046093.2 | NP_001039558.1 | 2 - 3 | 393 | 68 | |
GenBank mRNA | BC112594.1 | - | 2 - 3 | 394 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
