Assay Details
Gene Symbol: TDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046104.2 | NP_001039569.1 | 3 - 4 | 331 | 78 | |
GenBank mRNA | BC112535.1 | - | 3 - 4 | 332 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
