Assay Details
| Gene Symbol: DYDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046163.1 | NP_001039628.1 | 2 - 3 | 321 | 87 | |
| GenBank mRNA | BC112485.1 | - | 2 - 3 | 321 | 87 | |
| BC146140.1 | - | 4 - 5 | 690 | 87 | ||
More Information
Set Membership: |
|

DYDC2
Genomic Map