Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046220.2 | NP_001039685.1 | 1 - 2 | 150 | 65 | |
GenBank mRNA | BC110162.1 | - | 1 - 2 | 151 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110162.1, NM_001046220.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
