Assay Details
Gene Symbol: C18H19orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046227.2 | NP_001039692.2 | 3 - 4 | 135 | 83 | |
NM_001244559.1 | NP_001231488.1 | 3 - 4 | 129 | 83 | ||
GenBank mRNA | BC112702.1 | - | 3 - 4 | 101 | 83 | |
EV618076.1 | - | 3 - 4 | 128 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112702.1, NM_001046227.2, NM_001244559.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
