Assay Details
Gene Symbol: C18H19orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046227.2 | NP_001039692.2 | 4 - 5 | 174 | 74 | |
NM_001244559.1 | NP_001231488.1 | 4 - 5 | 168 | 74 | ||
GenBank mRNA | BC112702.1 | - | 4 - 5 | 140 | 74 | |
EV618076.1 | - | 4 - 5 | 167 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112702.1, NM_001046227.2, NM_001244559.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
