Assay Details
Gene Symbol: CALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046249.2 | NP_001039714.1 | 1 - 2 | 91 | 92 | |
GenBank mRNA | BC105380.1 | - | 1 - 2 | 92 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC105380.1, NM_001046249.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
