Assay Details
| Gene Symbol: TIMM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046254.1 | NP_001039719.1 | 4 - 5 | 473 | 95 | |
| GenBank mRNA | BC112498.1 | - | 4 - 5 | 473 | 95 | |
More Information
Set Membership: |
|

TIMM9
Genomic Map