Assay Details
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046286.2 | NP_001039751.1 | 6 - 7 | 960 | 131 | |
GenBank mRNA | BC112527.1 | - | 6 - 7 | 961 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
