Assay Details
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046316.2 | NP_001039781.1 | 5 - 6 | 671 | 67 | |
GenBank mRNA | BC105367.1 | - | 5 - 6 | 672 | 67 | |
BT030733.1 | - | 5 - 6 | 713 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
