Assay Details
| Gene Symbol: NT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046322.1 | NP_001039787.1 | 3 - 4 | 442 | 74 | |
| GenBank mRNA | BC110222.1 | - | 3 - 4 | 442 | 74 | |
More Information
Set Membership: |
|

NT5C
Genomic Map