Assay Details
| Gene Symbol: HADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046334.1 | NP_001039799.1 | 3 - 4 | 517 | 61 | |
| GenBank mRNA | BC105411.1 | - | 3 - 4 | 517 | 61 | |
More Information
Set Membership: |
|

HADH
Genomic Map