Assay Details
| Gene Symbol: COMMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046384.2 | NP_001039849.1 | 1 - 2 | 200 | 71 | |
| GenBank mRNA | BC111639.1 | - | 1 - 2 | 202 | 71 | |
More Information
Set Membership: |
|

COMMD1
Genomic Map