Assay Details
| Gene Symbol: HCFC1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046398.2 | NP_001039863.1 | 2 - 3 | 262 | 93 | |
| GenBank mRNA | BC111671.1 | - | 2 - 3 | 263 | 93 | |
| BC149392.1 | - | 1 - 2 | 1028 | 93 | ||
More Information
Set Membership: |
|

HCFC1R1
Genomic Map