Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046416.1 | NP_001039881.1 | 5 - 6 | 585 | 147 | |
| GenBank mRNA | BC110218.1 | - | 5 - 6 | 585 | 147 | |
More Information
Set Membership: |
|

AP3S1
Genomic Map